Skip to Content

C267

Monoclonal Anti-μ-Calpain (Domain II) antibody produced in mouse

clone 2H2A7C2, ascites fluid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
2H2A7C2, monoclonal
Application:
IF, WB
Citations:
6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2H2A7C2, monoclonal

mol wt

antigen 80 kDa

species reactivity

pig, rat, bovine, human

technique(s)

indirect immunofluorescence: 1:25, western blot: 1:1,000

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CAPN1(823)
rat ... Capn1(29153)

General description

Calpain-1 catalytic subunit is a protein encoded by the CAPN1 gene in humans. Calpains are calcium dependent proteases constituting a family of proteins. They share a homologous cysteine-protease domain and an E-F hand Ca2+-binding domain. The calpain system consists of two ubiquitous forms of calpain (m-calpain and μ-calpain), a tissue specific calpain (n-calpain) and a calpain inhibitory protein (calpastatin).

Immunogen

μ-calpain from bovine skeletal muscle.

Application

Monoclonal Anti-μ-Calpain (Domain II) antibody produced in mouse is suitable for indirect immunofluorescence at a dilution of 1:25 and western blotting at a dilution of 1:1000.

Biochem/physiol Actions

Calpain is involved in calmodulin-independent pathway for the activation of calcineurin. Endogenous calpain I forms active calcineurin in the human heart by proteolysis of calcineurin A, which may lead to pathogenesis of myocardial disease. μ-calpain, on overexpression, may have relationship with intractable epilepsy as well as the clinicopathological characteristics in such patients. Its activity may increase in skeletal muscle of gastric cancer patients.
Epitope mapping studies indicate the epitope is between amino acids 245-265 (domain II) of human μ-calpain. The antibody reacts specifically with μ-calpain. It does not cross-react with m-calpain, n-calpain, calmodulin or calpastatin. It is not recommended for immunoprecipitation. By immunoblotting, reactivity is observed with human platelets and erythrocytes, bovine platelets, heart and skeletal muscle and with rat myoblasts, kidney, liver and spleen. By immunofluorescence on pig LLC-PK1 cells, diffuse cytoplasmic staining is observed.

Physical form

Solution containing 0.05% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library