All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H9NO3
CAS Number:
Molecular Weight:
215.20
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
description
Light sensitive
Assay
≥90% (at 254 nm, HPLC)
form
powder
solubility
DMSO: soluble
fluorescence
λem 411 nm (+/- 4nm)
SMILES string
CCOc1ccc2C=C(C#N)C(=O)Oc2c1
InChI
1S/C12H9NO3/c1-2-15-10-4-3-8-5-9(7-13)12(14)16-11(8)6-10/h3-6H,2H2,1H3
InChI key
YAFGHMIAFYQSCF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Fluorescent probe useful in microsomal dealkylase studies.
Fluorescent probe useful in microsomal dealkylase studies. Typical drug-drug interactions resulting from enzyme inhibition.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chemopreventive and therapeutic potentials of thymoquinone in HepG2 cells: mechanistic perspectives.
Abeer ElKhoely et al.
Journal of natural medicines, 69(3), 313-323 (2015-03-23)
Liver cancer is the fifth commonest malignancy worldwide and the third leading cause of death. Identifying novel curative and preventive therapy may improve its prognosis. In this study, thymoquinone (TQ), the most active biological ingredient of Nigella sativa Linn, was
Kathleen W Mosure et al.
Journal of pharmaceutical sciences, 104(9), 2813-2823 (2015-01-30)
Asunaprevir (ASV; BMS-650032), a low nanomolar inhibitor of the hepatitis C virus (HCV) NS3 protease, is currently under development, in combination with other direct-acting antiviral (DAA) agents for the treatment of chronic HCV infection. Extensive nonclinical and pharmacokinetic studies have
Roslyn Thelingwani et al.
Expert opinion on drug metabolism & toxicology, 10(10), 1313-1324 (2014-09-16)
Drug resistance by Plasmodium falciparum remains a challenge in malaria chemotherapy. This paper will focus on physicochemical and drug metabolism characterization of a series of 4-aminoquinoline-3-hydroxypyridin-4-one hybrid shown to have antimalarial activity against chloroquine-resistant P. falciparum. The aim is to
Therese Ericsson et al.
Xenobiotica; the fate of foreign compounds in biological systems, 44(7), 615-626 (2014-01-10)
1. Cytochrome P450 enzyme system is the most important contributor to oxidative metabolism of drugs. Modification, and more specifically inhibition, of this system is an important determinant of several drug-drug interactions (DDIs). 2. Effects of the antimalarial agent artemisinin and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service