Skip to Content
Merck
All Photos(1)

Key Documents

C2363

Sigma-Aldrich

Monoclonal Anti-phospho-β-Catenin (pSer33) antibody produced in mouse

clone BC-76, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€508.00

€508.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
€508.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€508.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BC-76, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

microarray: suitable
western blot: 5 μg/mL using total cell extracts of cultured HEK-293T cells treated with MG132

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer33)

Gene Information

human ... CTNNB1(1499)

Related Categories

General description

Cateninβ is a multifunctional protein, coded by CTNNB1 (cateninβ 1) gene. It is present in the intracellular side of the cytoplasmic membrane. It is located on human chromosome 3p22.1.

Immunogen

synthetic phosphopeptide corresponding to the region that contains serine 33 of human β-catenin.

Application

Monoclonal Anti-phospho-β-Catenin (pSer33) antibody produced in mouse is suitable for microarray and western blotting at a concentration of 5μg/mL using total cell extracts of cultured HEK-293T cells treated with MG132.

Biochem/physiol Actions

Catenin (cadherin-associated protein), β 1 (88kDa) protein is encoded by CTNNB1 gene in humans. It is also called as β-catenin. It is a dual function protein that regulates the coordination of cell-cell adhesion and gene transcription. Human β-catenin is located at band p21 on chromosome 3.
Mutations in β-catenin results in retinal vascular condition FEVR (familial exudative vitreoretinopathy). It participates in stimulating disease progression, providing BCR–ABL (fusion gene) kinase-independent resistance and LSC (leukemic stem cell) self-renewal. CTNNB1 plays an important role in cell-to-cell adhesion and transcriptional modulation in the Wnt signalling pathway.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combined inhibition of β-catenin and Bcr?Abl synergistically targets tyrosine kinase inhibitor-resistant blast crisis chronic myeloid leukemia blasts and progenitors in vitro and in vivo.
Zhou H, et al.
Leukemia, 31(10), 2065?2074-2065?2074 (2017)
β-Catenin pathway activation in breast cancer is associated with triple-negative phenotype but not with CTNNB1 mutation.
Geyer FC, et al.
Modern Pathology, 24(2), 209?231-209?231 (2011)
P D McCrea et al.
Science (New York, N.Y.), 254(5036), 1359-1361 (1991-11-29)
Three cytoplasmic proteins, called catenins, bind to the cytoplasmic tail of the epithelial cell-cell adhesion molecule E-cadherin. The complementary DNA sequence was determined for the 92-kilodalton beta catenin of Xenopus laevis. The sequence is homologous to mammalian plakoglobin, a protein
Defects in the Cell Signaling Mediator ?-Catenin Cause the Retinal Vascular Condition FEVR
Panagiotou ES, et al.
American Journal of Human Genetics, 100(6), 960-968 (2017)
C Kraus et al.
Genomics, 23(1), 272-274 (1994-09-01)
The human beta-catenin locus (CTNNB1) was mapped by in situ fluorescence analysis to band p21 on the short arm of chromosome 3, a region frequently affected by somatic alterations in a variety of tumors. PCR primers for the genomic amplification

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service