Skip to Content
Merck
All Photos(1)

Key Documents

C2230

Sigma-Aldrich

Coelenterazine, native

solid

Synonym(s):

3,2-Dihydro-2-(p-hydroxybenzyl)-6-(p-hydroxyphenyl)-8-benzylimidazolo[1,2-a]pyrazin-3-one, CLZN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H21N3O3
CAS Number:
Molecular Weight:
423.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Assay

≥95% (TLC)

form

solid

solubility

methanol and ethanol: soluble

storage temp.

−20°C

SMILES string

Oc1ccc(CC2=NC3=C(Cc4ccccc4)NC(=CN3C2=O)c5ccc(O)cc5)cc1

InChI

1S/C26H21N3O3/c30-20-10-6-18(7-11-20)15-23-26(32)29-16-24(19-8-12-21(31)13-9-19)27-22(25(29)28-23)14-17-4-2-1-3-5-17/h1-13,16,27,30-31H,14-15H2

InChI key

YHIPILPTUVMWQT-UHFFFAOYSA-N

General description

Coelenterazine is a luciferin molecule, which is utilized by several luciferases, such as RenillaOplophorus and Periphylla.[1]

Application

Coelenterazine, native was used:-
  • For Aequorin assays[2]
  • For measuring the intracellular calcium concentration in response to ionomycin or PMA using aequorin in COS-7 cells[3]
  • In Bioluminescence imaging for analysing the presence of virus-dependent luciferase activity[4]

Biochem/physiol Actions

Luminophore of the aequorin complex used to measure Ca2+ concentration.
Luminophore of the aequorin complex which is oxidized by oxygen to illuminate at 465 nm when Ca2+ binds to the complex; used to measure Ca2+ concentration in cells with high sensitivity and large dynamic range.

Caution

Protect from exposure to light

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New bioluminescent coelenterazine derivatives with various C-6 substitutions
Jiang T, et al.
Organic & Biomolecular Chemistry, 15(33), 7008-7018 (2017)
Darren R Ritsick et al.
Free radical biology & medicine, 43(1), 31-38 (2007-06-15)
The catalytic subunit gp91phox (Nox2) of the NADPH oxidase of mammalian phagocytes is activated by microbes and immune mediators to produce large amounts of reactive oxygen species (ROS) which participate in microbial killing. Homologs of gp91phox, the Nox and Duox
Mengmeng Tang et al.
Plant methods, 16, 59-59 (2020-05-06)
The increase in atmospheric CO2 is causing a number of changes in plant growth such as increases in leaf area and number, branching, plant size and biomass, and growth rate. Despite the importance of stomatal responses to CO2, little is
I Gentschev et al.
Cancer gene therapy, 16(4), 320-328 (2008-10-25)
Mammary cancers together with cancers of the skin account for about 60% of the total cancers occurring in dogs. The veterinary options for therapeutic management of canine mammary cancer are limited and prognosis for such patients is poor. In this
Satoshi Inouye et al.
Analytical biochemistry, 407(2), 247-252 (2010-08-31)
Aequorin is a Ca(2+)-binding photoprotein and consists of an apoprotein (apoaequorin) and a 2-peroxide of coelenterazine. Eight new coelenterazine analogues modified at the C2-position were synthesized and incorporated into recombinant apoaequorin with O(2) to yield different semisynthetic aequorins. The luminescence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service