Skip to Content
Merck
All Photos(1)

Key Documents

C2149

Sigma-Aldrich

Cytochalasin E from Aspergillus clavatus

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H33NO7
CAS Number:
Molecular Weight:
495.56
Beilstein:
1096975
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

storage temp.

−20°C

SMILES string

C[C@H]1C\C=C\[C@H]2[C@@H]3O[C@]3(C)[C@@H](C)[C@H]4[C@H](Cc5ccccc5)NC(=O)[C@@]24OC(=O)O\C=C\[C@@](C)(O)C1=O

InChI

1S/C28H33NO7/c1-16-9-8-12-19-23-27(4,35-23)17(2)21-20(15-18-10-6-5-7-11-18)29-24(31)28(19,21)36-25(32)34-14-13-26(3,33)22(16)30/h5-8,10-14,16-17,19-21,23,33H,9,15H2,1-4H3,(H,29,31)/b12-8+,14-13+/t16-,17-,19-,20-,21-,23-,26+,27+,28+/m0/s1

InChI key

LAJXCUNOQSHRJO-ZYGJITOWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cytochalasin E has been used as:
  • a toxin to study its effects on avocado plants[1]
  • a component of the incubating medium in feline junctional adhesion molecule 1 (fJAM-1) expression assay[2]
  • an inhibitor of actin polymerization to study its effects on mitochondria uptake by mice endothelial cells[3]

Biochem/physiol Actions

Cytochalasin E is a cell-permeable fungal toxin that inhibits actin polymerization stimulated by F-actin. Cytochalasin E does not inhibit glucose transport.
Cytochalasin E is an epoxide[4] that exhibits anti-proliferative activity in endothelial cells in vitro. It also participates in inhibiting tumor growth and angiogenesis in vivo.[5] Cytochalasin E also possesses antimicrobial and antiviral properties.[4]

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contribution of Rosellinia necatrix toxins to avocado white root rot
Arjona-Girona I, et al.
European Journal of Plant Pathology, 148(1), 109-117 (2017)
Siwen Yuan et al.
European journal of medicinal chemistry, 202, 112502-112502 (2020-07-12)
Many fungal metabolites show promising anticancer properties both in vitro and in animal models, and some synthetic analogs of those metabolites have progressed into clinical trials. However, currently, there are still no fungi-derived agents approved as anticancer drugs. Two potential reasons
Liwen Lin et al.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 19(7), 1917-1929 (2019-02-15)
The innate immune system is a critical regulator of the adaptive immune responses that lead to allograft rejection. It is increasingly recognized that endogenous molecules released from tissue injury and cell death are potent activators of innate immunity. Mitochondria, ancestrally
Sylvie Delebassée et al.
Fitoterapia, 121, 146-151 (2017-07-15)
A biological screening of sixteen lichen extracts on human HT-29 colorectal cancer cells, led to the selection of Pleurosticta acetabulum, a lichen widely present in tree barks in Europe. Bioguided purification of the acetonic extract resulted in the isolation of
B O Gvakharia et al.
Cellular and molecular life sciences : CMLS, 60(8), 1744-1751 (2003-09-30)
Release of sperm bundles from moth testes is controlled by the local circadian oscillator. The mechanism which restricts migration of sperm bundles to a few hours each day is not understood. We demonstrate that a daily cycle of sperm release

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service