Skip to Content
Merck
All Photos(1)

Documents

C156

Sigma-Aldrich

D-CPT tartrate

>94%, solid

Synonym(s):

β-CPT tartrate, (–)-2β-Carbomethoxy-3β-phenyltropane tartrate, Troparil tartrate, WIN 35,065-2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21NO2 · C4H6O6
CAS Number:
Molecular Weight:
409.43
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

>94%

form

solid

optical activity

[α]22/D −95.9°, c = 1.1 in H2O(lit.)

drug control

USDEA Schedule II; Home Office Schedule 2; regulated under CDSA - not available from Sigma-Aldrich Canada

color

white

solubility

H2O: soluble
ethanol: soluble

storage temp.

−20°C

SMILES string

OC(C(O)C(O)=O)C(O)=O.COC(=O)[C@@H]1C2CCC(C[C@@H]1c3ccccc3)N2C

InChI

1S/C16H21NO2.C4H6O6/c1-17-12-8-9-14(17)15(16(18)19-2)13(10-12)11-6-4-3-5-7-11;5-1(3(7)8)2(6)4(9)10/h3-7,12-15H,8-10H2,1-2H3;1-2,5-6H,(H,7,8)(H,9,10)/t12-,13+,14+,15-;/m0./s1

InChI key

WYGLYLVLBCZESH-PEVLCXCCSA-N

Biochem/physiol Actions

Binds to the cocaine receptor site on the dopamine transporter and blocks dopamine uptake.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saturable (3H)cocaine binding in central nervous system of mouse.
M E Reith et al.
Life sciences, 27(12), 1055-1062 (1980-09-22)
J D Steketee
Behavioural brain research, 97(1-2), 39-48 (1998-12-29)
A role for the mesolimbic dopamine system in the development of behavioral sensitization to psychostimulants, such as cocaine and amphetamine, is well established. Previous reports have suggested that the ventral tegmental area (VTA) is involved in the initiation of, while
Emily M Jutkiewicz et al.
Annals of emergency medicine, 54(3), 409-420 (2008-11-18)
Cocaine toxicity results in cardiovascular complications, seizures, and death and accounts for approximately 20% of drug-related emergency department visits every year. Presently, there are no treatments to eliminate the toxic effects of cocaine. The present study hypothesizes that a bacterial
A Brouard et al.
Brain research. Developmental brain research, 75(1), 13-17 (1993-09-17)
The cellular localization of the cocaine binding sites in primary cultures of embryonic rat mesencephalic cells was previously reported to differ from that observed in adult rat brain. In order to know whether this different localization was associated with a
M E Reith
European journal of pharmacology, 130(1-2), 65-72 (1986-10-14)
Cocaine and a cocaine analog, WIN 36,065-2, were administered daily for 3 or 4 days. Both compounds developed sensitization to their locomotor stimulatory effects, with similarly shaped dose-response curves. Dosages giving optimal sensitization were 25 mg/kg per day for cocaine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service