Skip to Content
Merck
All Photos(1)

Key Documents

C-12980

Sigma-Aldrich

Human Pericytes from Placenta (hPC-PL)

500,000 cryopreserved cells

Synonym(s):

Pericyte Culture

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€1,530.00

€1,530.00


Estimated to ship on15 April 2025


Request a Bulk Order

Select a Size

Change View
1 EACH
€1,530.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

€1,530.00


Estimated to ship on15 April 2025


Request a Bulk Order

biological source

human placenta

packaging

pkg of 500,000 cells

morphology

( pericytes)

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Cell Line Origin

Placenta

Application

Human Pericytes are isolated from human placenta tissue and provided in a cryopreserved format. Pericytes are multipotent mesenchymal-like cells found in association with small blood vessel walls. They are important for angiogenesis, the structural integrity of the microvasculature, and blood flow regulation. However, they can also develop into malignant tumors called hemangiopericytomas. To date, pericytes have demonstrated the ability to differentiate into adipocytes, osteoblasts, fibroblasts, smooth muscle cells, and phagocytes (macrophages).

Quality

Rigid quality control tests are performed for each lot of Pericytes. The cells are tested for morphology, adherence rate, and viability. Furthermore, they are characterized by flow cytometric analysis for a comprehensive panel ofmarkers, e.g. CD31, CD34, CD105, and CD146. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Subculture Routine

Click here for more information.

Other Notes

Recommended Plating Density: 4000 cells per cm2Tested markers: CD146+, CD105+, CD31-, CD34-

Recommended products

Recommended Primary Cell Culture Media:Link

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

What are pericytes? What are common pericyte markers? Are pericytes mesenchymal stem cells? Learn more about pericytes in our FAQ page. Explore over 350 PromoCell products.

Primary human pericytes and cell culture media optimized for the in vitro cultivation of human pericytes from all tissues. Protocols for handling, cell passaging, media, and product use.

Smooth muscle human primary cells isolated from blood vessels and hollow organs, plus smooth muscle cell growth media. Protocols on media handling, cell passaging and product use.

Protocols

Cell culture protocol: the endothelial cell transwell migration and invasion assay used to study angiogenesis and cancer cell metastasis. Explore over 350 PromoCell products.

Related Content

Cell culture protocol: the endothelial tube formation assay to study angiogenesis using HUVECs and other endothelial cell types. Explore over 350 PromoCell products.

Questions

1–3 of 3 Questions  
  1. What markers are used to characterize pericytes?

    1 answer
    1. Pericytes are positive for CD146, PDGFRβ, αSMA, CD13, NG2, Desmin, and negative for CD31, CD34, CD45, and CD56.

      Helpful?

  2. How are pericytes isolated?

    1 answer
    1. Pericyte isolation generally starts with enzymatic digestion of tissue with Trypsin EDTA, usually followed by the isolation of microvessel fragments via successive filtration steps. Pure pericyte cultures can be obtained through either positive or negative immuno-selection via the use of magnetic beads or flow cytometry using the antibody markers above.

      Helpful?

  3. How long do pericytes grow in culture?

    1 answer
    1. PromoCell’s human pericytes derived from microvessels of the human placenta are cryopreserved at the end of secondary culture (P1) and after thawing are considered P2. The number of populations doublings is not determined for each individual cell lot, however they are guarenteed to be grown for at least 15 population doublings.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service