Skip to Content
Merck
All Photos(1)

Key Documents

C-12921

Sigma-Aldrich

Human CD34+ Progenitor Cells (hCD34+-CB)

Isolated from cord blood, single donor, 100,000 cryopreserved cells

Synonym(s):

hCD34+-CB cells

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€1,150.00

€1,150.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
€1,150.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

€1,150.00


Check Cart for Availability

Request a Bulk Order

biological source

human umbilical cord blood

packaging

pkg of 100,000 cells

morphology

(progenitor)

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Cell Line Origin

Cord Blood

Application

CD34 is a glycosylated transmembrane protein and represents a well-known marker for primitive blood- and bone marrow-derived progenitor cells, especially for hematopoietic and endothelial stem cells. Although the biological functions of CD34 are largely unknown, recent data suggest that CD34 is involved in maintenance of the progenitor cells in a phenotypically undifferentiated state. PromoCell offers CD34+ Progenitor Cells from the cord blood of healthy donors. The CD34+ Progenitor Cells contain two main cellular subpopulations, hematopoietic and endothelial progenitor cells. Therefore, CD34+ Progenitor Cells are suitable for a series of studies, e.g. directed differentiation into more committed types of blood cells and endothelial lineages.

Quality

Rigid quality control tests are performed for each lot of CD34+ Progenitor Cells. They are routinely characterized by flow cytometry analyzing the viability, cell size, and the presence of significant markers, i.e. CD34. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Subculture Routine

Click here for more information.

Other Notes

Recommended Plating Density: 5000 cells per mlTested Markers: CD34 positive

Recommended products

Recommended Primary Cell Culture Media:Link

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatjana Weidner et al.
Nature protocols, 16(7), 3210-3240 (2021-04-14)
Receptor targeting of vector particles is a key technology to enable cell type-specific in vivo gene delivery. For example, T cells in humanized mouse models can be modified by lentiviral vectors (LVs) targeted to human T-cell markers to enable them

Articles

CD34+ Progenitor Cells are suitable for diverse studies, as well as for the directed differentiation into more committed blood cell phenotypes and endothelial lineages. 

Yeast plasmid selection: Utilizes auxotrophic yeast strains for plasmid selection, enabling growth on synthetic media for better yield.

Overview of primary cells and media usage in biomedical culture applications highlights differences from cell lines.

Protocols

In vitro culture of human hematopoietic stem cells in PromoCell’s serum-free/xeno-free HPC Expansion Medium DXF. Explore over 350 PromoCell products

Hematopoietic stem cell culture protocols for isolation, expansion, and differentiation of CD34+ cells including CFU assays.

Generate mature human dendritic immune cells from PBMCs or CD34+ stem cell derived monocytes in 7 days in defined and serum-free dendritic cell culture media. Explore over 350 PromoCell products.

Questions

1–3 of 3 Questions  
  1. Do you have any information about the size of these cells? I need a human cell that can be cultured but less than 5 micrometer. Thanks.

    1 answer
    1. The cell size is not a defined release criterion for this product. However, literature indicates that Human CD34+ Progenitor Cells typically have diameters ranging from 7 to 10 micrometers.

      Helpful?

  2. Does it come from a single donor?

    1 answer
    1. Yes, as stated following the product name, these cells are isolated from a single donor.

      Helpful?

  3. I would like to know whether these are CD34+ progenitor cells isolated from cord blood and directly shipped after isolation or whether they are shipped after being cultured.

    1 answer
    1. Yes, these cells are CD34+ progenitor cells isolated from cord blood. PromoCell Blood and Blood Progenitor Cells are cryopreserved directly after isolation (=P0). Customers will receive them cryopreserved. They haven’t been in culture before freezing.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service