Skip to Content
Merck
All Photos(1)

Key Documents

C-12720

Sigma-Aldrich

Human Osteoblasts (HOB)

500,000 cryopreserved cells

Synonym(s):

HOB cells

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€1,120.00

€1,120.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
€1,120.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

€1,120.00


Check Cart for Availability

Request a Bulk Order

biological source

human bone (knee and hip joints)

packaging

pkg of 500,000 cells

morphology

(osteoblasts)

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Cell Line Origin

Bone Tissue

Application

Primary Human Osteoblasts (HOB) are isolated from femoral trabecular bone tissue from the knee or hip joint region (lot specific source information is available on request) and are provided in a cryopreserved format. They stain positive for alkaline phosphatase and can be mineralized using PromoCell Mineralization Medium.Osteoblasts are of mesenchymal origin and are mainly responsible for bone formation and remodeling. In vivo, they produce a collagen type I rich extracellular matrix called the osteoid. During their development, osteoblasts undergo a maturation process called osteogenesis. When they become encased in the bone matrix, they stop proliferating and become osteocytes.

Quality

Rigid quality control tests are performed for each lot of Human Osteoblasts. They are tested for cell morphology, adherence rate, and cell viability. Furthermore, histochemical tests for alkaline phosphatase and bone mineralization are carried out for each lot. Growth performance is tested through multiple passages up to 10 population doublings (PD) under culture conditions lacking antibiotics and antimycotics. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Subculture Routine

Click here for more information.

Other Notes

Recommended Plating Density: 10000 - 20000 cells per cm2Passage After Thawing: P2Tested Markers: Alkaline phosphatase positive, Mineralization testedGuaranteed population doublings: >10

Recommended products

Recommended Primary Cell Culture Media:Link

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilaria Lorenzon et al.
Cells, 9(1) (2019-12-28)
Platinum-based chemotherapy is the therapy of choice for epithelial ovarian cancer (EOC). Acquired resistance to platinum (PT) is a frequent event that leads to disease progression and predicts poor prognosis. To understand possible mechanisms underlying acquired PT-resistance, we have recently

Articles

Primary human osteoblasts and media for the optimal growth and mineralization of human osteoblast cultures. Protocols for handling, cell passaging, media, and product use.

Yeast plasmid selection: Utilizes auxotrophic yeast strains for plasmid selection, enabling growth on synthetic media for better yield.

Overview of primary cells and media usage in biomedical culture applications highlights differences from cell lines.

Related Content

Cell culture protocol used to differentiate primary human osteoblasts into bone cells in 2-3 weeks.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service