Select a Size
About This Item
Skip To
form
solid
Quality Level
optical activity
[α]25/D −62.5°, c = 1.0 in H2O(lit.)
drug control
USDEA Schedule II
color
white
solubility
H2O: soluble
storage temp.
2-8°C
SMILES string
O.OS(=O)(=O)c1cccc2c(cccc12)S(O)(=O)=O.COC(=O)[C@@H]3C4CCC(C[C@@H]3c5ccc(F)cc5)N4C
InChI
1S/C16H20FNO2.C10H8O6S2.H2O/c1-18-12-7-8-14(18)15(16(19)20-2)13(9-12)10-3-5-11(17)6-4-10;11-17(12,13)9-5-1-3-7-8(9)4-2-6-10(7)18(14,15)16;/h3-6,12-15H,7-9H2,1-2H3;1-6H,(H,11,12,13)(H,14,15,16);1H2/t12-,13+,14+,15-;;/m0../s1
InChI key
XSJPWVYASBDEJI-YEVYTSHMSA-N
Biochem/physiol Actions
Features and Benefits
signalword
Danger
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Repr. 2 - Skin Sens. 1
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk
WGK 3
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C124-5MG | 04061832089560 |
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service
