Skip to Content
Merck
All Photos(1)

Key Documents

C1116

Sigma-Aldrich

Anti-Chloride Channel CLC-5 (Clcn5) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-CLC5, Anti-CLCK2, Anti-ClC-5, Anti-DENT1, Anti-DENTS, Anti-NPHL1, Anti-NPHL2, Anti-XLRH, Anti-XRN

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€291.00
200 μL
€826.00

€291.00


Check Cart for Availability


Select a Size

Change View
50 μL
€291.00
200 μL
€826.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€291.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human, mouse, rat

technique(s)

western blot: 1:200 using rat kidney membranes

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLCN5(1184)
mouse ... Clcn5(12728)
rat ... Clcn5(25749)

Immunogen

peptide corresponding to amino acid residues 401-415 of rat CLC5. Mouse sequence is identical; human sequence is 14/15 residues identical.

Application

Anti-Chloride Channel CLC-5 (Clcn5) antibody produced in rabbit is suitable for western blotting at a dilution of 1:200 using rat kidney membranes.

Biochem/physiol Actions

H(+)/Cl(-) exchange transporter 5 is a protein encoded by the CLCN5 gene in humans. It encodes a member of the CLC gene family of chloride ion channels and ion transporters. CLCN5 is highly expressed in endosomes of proximal tubule cells and is essential for endocytosis. Mutations in CLCN5 causes Dent′s disease leading to renal failure. It is also involved in low-molecular-weight proteinuria, hypercalciuria, nephrolithiasis and renal failure. Majority of these disease-causing mutations in ClC-5 are misprocessed and retained in the ER (endoplasmic reticulum) and may alter intramolecular interactions within the full-length ClC-5 protein. CLC-5 plays a crucial role in the process of endocytosis in the proximal tubule of the kidney and mutations that alter protein function are the cause of Dent′s disease. It may act as an electrically shunting Cl- channel in early endosomes, facilitating intraluminal acidification.

Physical form

Lyophilized from phosphate buffered saline containing, pH 7.4, 1% BSA and 0.05% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia De Stefano et al.
The Journal of physiology, 591(23), 5879-5893 (2013-10-09)
ClC-5 is a 2Cl(-)/1H(+) antiporter highly expressed in endosomes of proximal tubule cells. It is essential for endocytosis and mutations in ClC-5 cause Dent's disease, potentially leading to renal failure. However, the physiological role of ClC-5 is still unclear. One
Miriam F Figueira et al.
Physiological reports, 5(13) (2017-07-06)
Diabetic nephropathy (DN) occurs in around 40% of those with diabetes. Proteinuria is the main characteristic of DN and develops as a result of increased permeability of the glomerulus capillary wall and/or decreased proximal tubule endocytosis. The goal of this
Hengli Zhang et al.
Clinical & experimental optometry, 94(6), 528-535 (2011-09-08)
Experimental evidence has shown that myopic and hyperopic optical defocus induces thickening and thinning of the choroids, respectively, moving the retina forward and backward toward the plane of focus; however, the underlying mechanism of this phenomenon remains elusive. It has
Christina D'Antonio et al.
The Biochemical journal, 452(3), 391-400 (2013-04-10)
Mutations in the CLCN5 (chloride channel, voltage-sensitive 5) gene cause Dent's disease because they reduce the functional expression of the ClC-5 chloride/proton transporter in the recycling endosomes of proximal tubule epithelial cells. The majority (60%) of these disease-causing mutations in
Jonathan D Lippiat et al.
Frontiers in physiology, 3, 449-449 (2012-12-12)
CLC-5 plays a critical role in the process of endocytosis in the proximal tubule of the kidney and mutations that alter protein function are the cause of Dent's I disease. In this X-linked disorder impaired reabsorption results in the wasting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service