Skip to Content
Merck
All Photos(1)

Key Documents

C1011

Sigma-Aldrich

N-Cyclohexyl-N′-(2-morpholinoethyl)carbodiimide metho-p-toluenesulfonate

≥97% (CHN)

Synonym(s):

N-Cyclohexyl-N′-(β-[N-methylmorpholino]ethyl)carbodiimide p-toluenesulfonate, CMC, CME-CDI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H26N3O · C7H7O3S
CAS Number:
Molecular Weight:
423.57
Beilstein:
3872192
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (CHN)

form

powder

technique(s)

ligand binding assay: suitable

color

white

mp

109-111 °C

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

Cc1ccc(cc1)S([O-])(=O)=O.C[N+]2(CCOCC2)CCN=C=NC3CCCCC3

InChI

1S/C14H26N3O.C7H8O3S/c1-17(9-11-18-12-10-17)8-7-15-13-16-14-5-3-2-4-6-14;1-6-2-4-7(5-3-6)11(8,9)10/h14H,2-12H2,1H3;2-5H,1H3,(H,8,9,10)/q+1;/p-1

InChI key

GBCAVSYHPPARHX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Water-soluble condensing reagent for peptide synthesis, reagent used for coupling ligands to carriers in affinity chromatography.

Biochem/physiol Actions

N-Cyclohexyl-N′-(2-morpholinoethyl)carbodiimide (CMCT) selectively modifies/derivitizes all pseudouridine, thiouridine and 2-methylthio-6-isopentenyladenosine nucleosides for detection by methods such as matrix-assisted laser desorption/ionization mass spectrometry. CMCT, a water-soluble condensing reagent for peptide synthesis, is used as a reagent for coupling ligands to carriers in affinity chromatography.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mei-Hui Wang et al.
Journal of hepatology, 63(2), 370-377 (2015-03-24)
The asialoglycoprotein receptor on hepatocyte membranes recognizes the galactose residues of glycoproteins. We investigated the specificity, accuracy and threshold value of asialoglycoprotein receptor imaging for estimating liver reserve via scintigraphy using (111)In-hexavalent lactoside in mouse models. (111)In-hexavalent lactoside scintigraphy for
M S Sloma et al.
Biochimica et biophysica acta, 1521(1-3), 30-38 (2001-11-03)
We have investigated possible interaction sites for mRNA, tRNA, translation factors and the nascent peptide on 5S, 5.8S and 28S rRNA in in vivo assembled translational active mouse ribosomes by comparing the chemical footprinting patterns derived from native polysomes, salt-washed
Yubiao Liu et al.
Chemical communications (Cambridge, England), (4)(4), 398-399 (2004-02-07)
Poly(N,N-dialkylcarbodiimide) was found to be an effective polymeric ligand system for preparing and stabilizing palladium nanoparticles (1-5 nm). The composite material prepared in situ was found to be a robust catalyst for the Suzuki coupling reaction under microwave or regular
Katherine A Kellersberger et al.
Analytical chemistry, 76(9), 2438-2445 (2004-05-01)
A top-down approach based on sustained off-resonance irradiation collision-induced dissociation (SORI-CID) has been implemented on an electrospray ionization (ESI) Fourier transform mass spectrometer (FTMS) to characterize nucleic acid substrates modified by structural probes. Solvent accessibility reagents, such as dimethyl sulfate
Yongjun Wei et al.
PloS one, 10(6), e0129921-e0129921 (2015-06-13)
Diverse cellulolytic bacteria are essential for maintaining high lignocellulose degradation ability in biogas digesters. However, little was known about functional genes and gene clusters of dominant cellulolytic bacteria in biogas digesters. This is the foundation to understand lignocellulose degradation mechanisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service