Skip to Content
Merck
All Photos(2)

Documents

C0987

Sigma-Aldrich

CGP-13501

≥98% (HPLC), solid

Synonym(s):

3-(3′,5′-Di-tert-butyl-4′-hydroxy)phenyl-2,2-dimethylpropanal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O2
CAS Number:
Molecular Weight:
290.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: soluble 24 mg/mL

originator

Novartis

SMILES string

[H]C(=O)C(C)(C)Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C19H30O2/c1-17(2,3)14-9-13(11-19(7,8)12-20)10-15(16(14)21)18(4,5)6/h9-10,12,21H,11H2,1-8H3

InChI key

XGWATTXMMMANFJ-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Positive allosteric modulator of GABAB receptors.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GABAB Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service