Skip to Content
Merck

Skip To

C0817

Monoclonal Anti-HLA-DQ antibody produced in mouse

clone HK19, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-HLA-DQ

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
HK19, monoclonal
Application:
FACS
Citations:
3


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HK19, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: 5 μL using 1 × 106 cells

isotype

IgG2b

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... HLA-DQB1(3119)

General description

HLA-DQ molecules are bimolecular glycoprotein complexes of 29 kDa and 33 kDa components. The HLA-DQ antigens are present on peripheral blood lymphocytes and tonsillar and splenic mononuclear cells.

Immunogen

GM-1500 human tumor cell line.

Application

Monoclonal Anti-HLA-DQ antibody produced in mouse is suitable for flow cytometry at a concentration of 5 μL using 1 × 106 cells.

Biochem/physiol Actions

Human leukocyte antigen (HLA)-DQ is a cell surface receptor type protein found on antigen presenting cells. DQ is a αβ heterodimer of the MHC Class II type. It plays a central role in the immune system and is expressed in antigen presenting cells. HLA-DQ expression in cystic fibrosis CF is associated with inflammation and reduces response to Interferon γ (IFNγ). HLA-DQ acts as alloantibody most frequently associated with the generation of de novo donor-specific antibody (DSA) and antibody-mediated-rejection but does not support organ transplantation.
Recognizes MHC Class II molecules that are preferentially expressed on the surface of B lymphocytes. The antibody reacts weakly with resting or ConA-activated T cells, monocytes, granulocytes, and erythrocytes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters