Key Documents
BMSIPD4400
Sigma-Aldrich
Benchmark BenchMasher
AC/DC input 120 V
Synonym(s):
masher, bag homogeniser, bag blender
About This Item
UNSPSC Code:
41103800
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPD4400
parameter
3-12 speed (strokes per sec)
General description
The BenchMasher provides fast and effective mixing, ensuring consistent homogenization. It is ideal for preparation of samples were eliminating contamination is critical, such as pathogen and microbial detection in food and water filters. The BenchMasher also has applications anywhere larger solid samples need broken down or products such as powders and polymers need mixed.Samples up to 400ml are conveniently loaded in the sterile blender bags which are sealed upon closing the door. Mixing paddles rapidly strike the bags, producing thorough mixing and homogenization without ever directly contacting the sample. Programming is user friendly – mixing speed and cycle time are easily adjusted on the large, full color touch screen. Paddle position can be adjusted to ensure efficient processing of samples. For applications requiring thermal regulation (such as Trichinella testing in meat) the chamber can be heated and maintained at temperatures up to 65°C.The door is removable for access to the chamber and stainless steel mixing paddles. Each paddle includes a convenient rubber cover that is easily removed or replaced. A UV light (254nm) is built into the chamber and can be activated at any time to perform complete decontamination.
Features and Benefits
  • Contamination free mixing/homogenizing of samples
  • Touch screen control with variable mixing speed and temperature control
  • Adjustable stroke length to accommodate samples of various types
  • Window for sample visualization
  • Built in UV light for chamber decontamination