Skip to Content
Merck
All Photos(1)

Key Documents

BM0009

Sigma-Aldrich

Avitriptan fumarate

≥98% (HPLC)

Synonym(s):

3-[3-[4-(5-Methoxy-4-pyrimidinyl)-1-piperazinyl]propyl]-N-methyl-1H-indole-5-methanesulfonamide (E)-2-butenedioate (1:1), BMS-180048, BMS180048

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30N6O3S · C4H4O4
CAS Number:
Molecular Weight:
574.65
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

InChI

1S/C22H30N6O3S.C4H4O4/c1-23-32(29,30)15-17-5-6-20-19(12-17)18(13-25-20)4-3-7-27-8-10-28(11-9-27)22-21(31-2)14-24-16-26-22;5-3(6)1-2-4(7)8/h5-6,12-14,16,23,25H,3-4,7-11,15H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1+

InChI key

FMXXORRZIQGUIN-WLHGVMLRSA-N

Biochem/physiol Actions

Avitriptan fumarate is a serotonin 5HT-1B/1D-receptor agonist with anti-migraine activity.
Avitriptan fumarate is a serotonin 5HT-1B/1D-receptor agonist.

Features and Benefits

Avitriptan is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbora Vyhlídalová et al.
International journal of molecular sciences, 21(8) (2020-04-23)
The efforts for therapeutic targeting of the aryl hydrocarbon receptor (AhR) have emerged in recent years. We investigated the effects of available antimigraine triptan drugs, having an indole core in their structure, on AhR signaling in human hepatic and intestinal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service