Skip to Content
Merck
All Photos(1)

Documents

B9258

Sigma-Aldrich

2-Bromoethylamine hydrobromide

Synonym(s):

2-Aminoethyl bromide hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2NH2 · HBr
CAS Number:
Molecular Weight:
204.89
Beilstein:
3607605
EC Number:
MDL number:
UNSPSC Code:
12352200

mp

170-175 °C (lit.)

SMILES string

Br.NCCBr

InChI

1S/C2H6BrN.BrH/c3-1-2-4;/h1-2,4H2;1H

InChI key

WJAXXWSZNSFVNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sai An et al.
Small (Weinheim an der Bergstrasse, Germany), 15(9), e1805182-e1805182 (2019-01-29)
Triple negative breast cancer (TNBC) is the most aggressive breast cancer subtype. Currently, no targeted treatment is available for TNBC, and the most common clinical therapy is tumor resection, which often promotes metastasis risks. Strong evidence suggests that the lymphatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service