Skip to Content
Merck
All Photos(1)

Key Documents

B9186

Sigma-Aldrich

Anti-Brain-derived Neurotrophic Factor antibody produced in chicken

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-BDNF, Anti-MGC34632

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

flow cytometry: 3-10 μg/mL
immunohistochemistry: 0.1-1.0 μg/mL using free-floating sections
immunohistochemistry: 0.5-5 μg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... BDNF(627)

Immunogen

purified, Sf21-derived, recombinant human brain-derived neurotrophic factor.

Biochem/physiol Actions

Brain derived neurotrophic factor (BDNF) belongs to a family of secreted proteins called neurotrophins that includes, nerve growth factor (NGF), neurotrophin-3 (NT3) and NT4/5. BDNF is expressed in the developing and the mature brain. The downstream pathways promoted by BDNF include PI3K, MAPK and JAK/STAT. The most important role of BDNF is the regulation of synaptic transmission and plasticity, protein synthesis and phosphorylation of local proteins.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hai-Yang Zhang et al.
Asian journal of andrology, 13(2), 231-235 (2010-12-21)
Retroperitoneal operations, such as radical prostatectomy, often damage the cavernous nerve, resulting in a high incidence of erectile dysfunction. Although improved nerve-sparing techniques have reduced the incidence of nerve injury, and the administration of phosphodiesterase type 5 inhibitors has revolutionized
Susana Cohen-Cory et al.
Developmental neurobiology, 70(5), 271-288 (2010-02-27)
During development, neural networks are established in a highly organized manner, which persists throughout life. Neurotrophins play crucial roles in the developing nervous system. Among the neurotrophins, brain-derived neurotrophic factor (BDNF) is highly conserved in gene structure and function during
Emily G Waterhouse et al.
Molecular and cellular neurosciences, 42(2), 81-89 (2009-07-07)
Substantial evidence indicates that brain-derived neurotrophic factor (BDNF) plays a crucial role in synaptic plasticity. Long-lasting synaptic plasticity is restricted to active synapses and requires new protein synthesis. Recent work has identified local protein synthesis as an important source for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service