Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Check Cart for Availability | € 37,60 |
About This Item
NACRES:
NB.35
UNSPSC Code:
41122800
Material:
heavy wire frame
€ 37,60
Check Cart for Availability
material
heavy wire frame
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H13193-1000
L × W × H
12.5 cm (5 in.) × 7.5 cm (3 in.) × 20 cm (8 in.)
General description
Holds 22 × 28 cm bags open for disposal of small items like microcentrifuge tubes and pipet tips.
1 of 1
This Item | |||
|---|---|---|---|
| material heavy wire frame | material - | material - | material - |
| L × W × H 12.5 cm (5 in.) × 7.5 cm (3 in.) × 20 cm (8 in.) | L × W × H 33 cm (13 in.) × 35 cm (13 3/4 in.) × 72 cm (28 in.) | L × W × H - | L × W × H - |
| manufacturer/tradename Bel-Art H13193-1000 | manufacturer/tradename Bel-Art F13192-0003 | manufacturer/tradename Bel-Art F13164-2535 | manufacturer/tradename Bel-Art F13166-0000 |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 200 | packaging pack of 100 |
Still not finding the right product?
Explore all of our products under Biohazard disposal stand
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


