The approximate amount of known components are listed below:
8.0 - 9.5% cholic acid
0.9 - 1.1% deoxycholic acid
32 - 36% conjugated cholic acid
11 - 13% conjugated deoxycholic acid
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
10 g | Check Cart for Availability | € 49,60 |
25 g | Check Cart for Availability | € 81,30 |
100 g | Check Cart for Availability | € 275,00 |
About This Item
Form:
powder
Biological source:
bovine, ovine
€ 49,60
Check Cart for Availability
Quality Segment
biological source
bovine, ovine
form
powder
technique(s)
microbiological culture: suitable
Application
Bile from bovine and ovine can be used in bacteriological research to investigate bile salt adaptation of bacteria such as Lactobacillus.
For bacteriological use.
Biochem/physiol Actions
The formation of bile acids from cholesterol is critical to cholesterol homeostasis. Subsequently, bile acids and alcohols function in the absorption of lipids from the GI tract. The class consists of a complex mixture of similar compounds, presenting a difficult analytical problem; LC/MS and GC/MS have been used successfully to elucidate such mixtures.
Other Notes
Mixture of free and conjugated bile acids, minimum 50% bile acids
1 of 1
This Item | |||
|---|---|---|---|
| description bile acid mixture | description - | description - | description heat shock fraction, low endotoxin, pH 7, ≥98% |
| biological source bovine | biological source Porcine | biological source Porcine, bovine | biological source bovine |
| technique(s) microbiological culture: suitable | technique(s) microbiological culture: suitable | technique(s) microbiological culture: suitable | technique(s) microbiological culture: suitable, ELISA: suitable |
| form powder | form powder | form powder | form lyophilized powder |
| Quality Level 200 | Quality Level 200 | Quality Level 100 | Quality Level 200 |
Hazard Codes
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class
11 - Combustible Solids
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Could you disclose the composition of this product (the bile acids themselves)? And its percentages.
1 answer-
Helpful?
-



