Skip to Content
Merck
All Photos(1)

Key Documents

B8131

Sigma-Aldrich

2-(2′-Benzothiazolyl)-5-styryl-3-(4′-phthalhydrazidyl)tetrazolium chloride

90-95%

Synonym(s):

BPST Chloride, BSPT Chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16ClN7O2S
CAS Number:
Molecular Weight:
501.95
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

90-95%

storage temp.

2-8°C

SMILES string

[Cl-].O=C1NNC(=O)c2cc(ccc12)-[n+]3nc(\C=C\c4ccccc4)nn3-c5nc6ccccc6s5

InChI

1S/C24H15N7O2S.ClH/c32-22-17-12-11-16(14-18(17)23(33)27-26-22)30-28-21(13-10-15-6-2-1-3-7-15)29-31(30)24-25-19-8-4-5-9-20(19)34-24;/h1-14H,(H,28,29);1H/b13-10+;

InChI key

ZPGCKYFKHCIWHR-RSGUCCNWSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of BSPT in dehydrogenase histochemistry.
W A Shannon
The Histochemical journal, 14(1), 166-168 (1982-01-01)
G Wolf et al.
Neuroscience letters, 161(1), 49-52 (1993-10-14)
The 2-(2'-benzothiazolyl)-5-styryl-3-(4'-phthalhydrazidyl) tetrazolium chloride (BSPT)-tetrazolium salt technique for the electron microscopic demonstration of reduced nicotinamide adenine dinucleotide phosphate-diaphorase (NADPH-d) was used to localize nitric oxide synthase in the normal and excitotoxically lesioned rat hippocampus. The reaction product BSPT-formazan was shown
S Barbey et al.
Pathology, research and practice, 182(6), 805-809 (1987-12-01)
The ultrastructural and histochemical exploration of histiocytes X done by the means of Concanavalin A (Con A), peanut agglutinin (PNA) and BSPT demonstrate that the plasma membrane of histiocytes X shared some properties with all the other cells and is
S Darius et al.
Brain research, 690(2), 231-235 (1995-09-04)
The activity of NADPH-diaphorase (NADPH-d), a marker for nitric oxide synthase (NOS), was examined histochemically in the rat and mice retina. Mice in which the neuronal NOS gene has been disrupted (nNOS- mice) were used for specificity controls. Light microscopically
S Matsubara et al.
European journal of histochemistry : EJH, 43(3), 247-251 (1999-11-24)
We investigated the subcellular localization of nicotinamide adenine dinucleotide phosphate diaphorase (NADPH-d) activity, a histo- and cyto-chemical marker of nitric oxide synthase, in human placental trophoblast obtained from women with normal term pregnancies. Tetrazolium salt BSPT was used as the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service