Skip to Content
Merck

Skip To

B7313

L-BNP trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

Ser-Pro-Lys-Met-Val-Gln-Gly-Ser-Gly-Cys-Phe-Gly-Leu-Lys-Met-Asp-Arg-Ile-Ser-Ser-Ser-Ser-Gly-leu-Gly-Cys-Lys-Val-Leu-Arg-Arg-His [Disulfide Bridge: 10-26] trifluoroacetate salt, [Leu13]-BNP trifluoroacetate salt, [Leu13]-Brain Natriuretic Peptide-32 trifluoroacetate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 488,00

About This Item

Empirical Formula (Hill Notation):
C143H243N47O42S4 · xC2HF3O2
Molecular Weight:
3421.01 (free base basis)
NACRES:
NA.32
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder

€ 488,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to off-white

storage temp.

−20°C

Biochem/physiol Actions

L-BNP (Arg13Leu-BNP) is a substituted Brain Natiuretic Peptide with discriminate guanylyl cyclase activator.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2988SML3198SML2627
assay

≥98% (HPLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

≥95% (HPLC)

form

powder

form

powder

form

powder

form

lyophilized powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

color

white to off-white

color

white to off-white

color

white to off-white

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters