Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
25 ea | Check Cart for Availability | € 46,40 |
About This Item
€ 46,40
material
colorless bottle, low-density polyethylene , polypropylene cap, round bottle
joint
Joints threaded neck (15)
packaging
pack of 25 ea
manufacturer/tradename
Nalgene® 2750-9050
capacity
15 mL
color
white cap
Features and Benefits
- Nalgene® dropper bottles with screw closure is an alternative to various pipetting devices.
- The tip ensures accurate dispensing of 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time.
- Chemically resistant to most reagents.
- LDPE clear transparent bottles for easy sample visualization.
- Leak-proof closure - prevents sample loss.
- Control dispensing tip ensures a secure fit.
- Suitable for use in biotech, diagnostic and pharmaceutical applications.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| material colorless bottle, polypropylene cap, low-density polyethylene , round bottle | material low-density polyethylene , natural bottle, round bottle | material low-density polyethylene , round bottle, white bottle | material low-density polyethylene , white bottle |
| manufacturer/tradename Nalgene® 2750-9050 | manufacturer/tradename Nalgene® 2752-9050 | manufacturer/tradename Nalgene® 2753-9050 | manufacturer/tradename Nalgene® 2751-9050 |
| capacity 15 mL | capacity 15 mL | capacity 15 mL | capacity 15 mL |
| joint Joints threaded neck (15) | joint Joints threaded neck (15) | joint Joints threaded neck (15-415) | joint Joints threaded neck (15-415) |
| packaging pack of 25 ea | packaging pack of 25 ea | packaging pack of 25 ea | packaging pack of 25 ea |
| color white cap | color assorted colors cap | color assorted colors cap | color white cap |
Still not finding the right product?
Explore all of our products under Nalgene® dropper bottle with screw closure
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
