B5920
6-Benzylaminopurine hydrochloride
suitable for plant cell culture, ≥98%
Synonym(s):
BA, N6-Benzyladenine
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Empirical Formula (Hill Notation):
C12H11N5 · HCl
CAS Number:
Molecular Weight:
261.71
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98%
form:
powder
solubility:
H2O: soluble
Recommended Products
Assay
≥98%
form
powder
technique(s)
cell culture | plant: suitable
solubility
H2O: soluble
application(s)
agriculture
SMILES string
Cl.C(Nc1ncnc2nc[nH]c12)c3ccccc3
InChI
1S/C12H11N5.ClH/c1-2-4-9(5-3-1)6-13-11-10-12(15-7-14-10)17-8-16-11;/h1-5,7-8H,6H2,(H2,13,14,15,16,17);1H
InChI key
VQVCNMLGIVVDOS-UHFFFAOYSA-N
General description
6-benzylaminopurine (6-BAP), a synthetic phytohormone, is a first-generation pesticide.
Biochem/physiol Actions
6-Benzylaminopurine (6-BAP), a cytokinin, can promote the growth and development of plants. It helps to set blossoms, initiate fruit maturity and blocks respiratory kinase in plants. 6-BAP is capable of enhancing post harvest life of green vegetables.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Runlong Hao et al.
Environmental science & technology, 53(15), 9014-9023 (2019-07-03)
A novel advanced oxidation process (AOP) using ultraviolet/sodium chlorite (UV/NaClO2) is developed for simultaneous removal of SO2 and NO. NH4OH, as an additive, was used to inhibit the generation of ClO2 and NO2. The removal efficiencies of SO2 and NO
Ayesha Sengupta et al.
Neuron, 103(3), 489-505 (2019-06-18)
Despite a wealth of clinical and preclinical data implicating the serotonin (5-HT) system in fear-related affective disorders, a precise definition of this neuromodulator's role in fear remains elusive. Using convergent anatomical and functional approaches, we interrogate the contribution to fear
Julie Samora et al.
The Journal of the American Academy of Orthopaedic Surgeons, 27(8), e360-e363 (2018-10-10)
The American Academy of Orthopaedic Surgeons has developed the Appropriate Use Criteria (AUC) for the Management of Developmental Dysplasia of the Hip in Infants up to Six Months of Age: Intended for Use by Orthopaedic Specialists. Evidence-based information, in conjunction
Haizhen Yang et al.
ACS applied materials & interfaces, 11(32), 29218-29230 (2019-07-20)
The porosity, roughness, and thickness of woven fabrics limit inkjet printing quality, which is extremely important for obtaining high-quality inkjet printing images on fabrics. This study reveals the application of poly[styrene-butyl acrylate-(P-vinylbenzyl trimethyl ammonium chloride)] nanospheres prepared via a soap-free
Galina Y Zheleznyakova et al.
Physiological genomics, 49(9), 447-461 (2017-07-30)
Multiple sclerosis (MS) is a chronic inflammatory and demyelinating disease of the central nervous system. MS likely results from a complex interplay between predisposing causal gene variants (the strongest influence coming from HLA class II locus) and environmental risk factors
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service