B5763
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide
Synonym(s):
MTBSTFA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CF3CON(CH3)Si(CH3)2C(CH3)3
CAS Number:
Molecular Weight:
241.33
Beilstein:
3606546
MDL number:
UNSPSC Code:
12352000
refractive index
n20/D 1.402 (lit.)
bp
172-175 °C (lit.)
density
1.036 g/mL at 25 °C (lit.)
storage temp.
−20°C
SMILES string
CN(C(=O)C(F)(F)F)[Si](C)(C)C(C)(C)C
InChI
1S/C9H18F3NOSi/c1-8(2,3)15(5,6)13(4)7(14)9(10,11)12/h1-6H3
InChI key
QRKUHYFDBWGLHJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Useful silylating reagent, less moisture sensitive than others.
replaced by
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
113.0 °F - closed cup
Flash Point(C)
45 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
P Canosa et al.
Journal of chromatography. A, 1072(1), 107-115 (2005-05-11)
A solid-phase microextraction (SPME) method for the determination of triclosan, methyl triclosan, 2,4-dichlorophenol and 2,3,4-trichlorophenol (considered as possible triclosan metabolites) in water samples was optimised. Analytes were first concentrated on a SPME fibre, directly exposed to the sample, and then
Xiaodong Huang et al.
Analytical chemistry, 80(1), 107-114 (2007-12-07)
This work describes an approach to differential metabolomics that involves stable isotope labeling for relative quantification as part of sample analysis by two-dimensional gas chromatography/mass spectrometry (GCxGC/MS). The polar metabolome in control and experimental samples was extracted and differentially derivatized
Claude Schummer et al.
Talanta, 77(4), 1473-1482 (2008-12-17)
In this study, MTBSTFA and BSTFA, which are among the preferred derivatization reagents for silylation were both tested on derivatization of six different groups of polar chemicals to get information about usefulness in terms of sensitivity and specificity of both
Paul L Wood et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 831(1-2), 313-319 (2006-01-13)
The GC-MS quantitation of a large number of neurochemicals utilizing a single derivatization step is not common but is provided by the reagent N-(tert-butyldimethylsilyl)-N-methyltrifluro-acetamide (MTBSTFA). Previous workers have utilized this derivative for GC-MS analyses of amino acids, carboxylic acids and
Martin Adiels et al.
Rapid communications in mass spectrometry : RCM, 24(5), 586-592 (2010-02-09)
Stable isotope kinetic studies play an important role in the study of very-low density lipoprotein (VLDL) metabolism, including basic and clinical research. Today, [1,1,2,3,3-(2)H(5)]glycerol is the most cost-effective alternative to measure glycerol and triglyceride kinetics. Recycling of glycerol from glycolysis
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service