Skip to Content
Merck

Skip To

B5313

BIX-01338 hydrate

≥98% (HPLC)

Synonym(s):

1-{2-[4-(4-Methoxybenzoyloxy)phenyl]ethyl}-2-(4-trifluoromethylbenzoylamino)-1H-benzoimidazole-5-carboxylic acid hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 199,00
€ 169,15
25 mg
Check Cart for Availability
€ 633,00
€ 538,05

About This Item

Empirical Formula (Hill Notation):
C32H24F3N3O6 · xH2O
CAS Number:
Molecular Weight:
603.54 (anhydrous basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder

€ 169,15

List Price€ 199,00Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

off-white to light tan

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

O.COc1ccc(cc1)C(=O)Oc2ccc(CCn3c(NC(=O)c4ccc(cc4)C(F)(F)F)nc5cc(ccc35)C(O)=O)cc2

InChI

1S/C32H24F3N3O6.H2O/c1-43-24-13-6-21(7-14-24)30(42)44-25-11-2-19(3-12-25)16-17-38-27-15-8-22(29(40)41)18-26(27)36-31(38)37-28(39)20-4-9-23(10-5-20)32(33,34)35;/h2-15,18H,16-17H2,1H3,(H,40,41)(H,36,37,39);1H2

InChI key

LLTPCGBKXOESTJ-UHFFFAOYSA-N

Biochem/physiol Actions

BIX-01338 is a non-selective histone lysine methyltransferase inhibitor.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G5793S7076C3742
form

powder

form

powder

form

solid

form

solid

assay

≥98% (HPLC)

assay

>98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: >10 mg/mL

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: ≥12 mg/mL

solubility

DMSO: ≥2 mg/mL (warmed)

color

off-white to light tan

color

white to beige

color

white to beige

color

off-white to tan


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters