Skip to Content
Merck
All Photos(1)

Key Documents

B5277

Sigma-Aldrich

Boc-Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Ile-Arg-Pro-Arg-Leu-Arg-Gly 5-aminopentylamide

≥97% (HPLC)

Synonym(s):

Boc-DAKLI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C87H149N31O17
CAS Number:
Molecular Weight:
1901.31
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

composition

Peptide content, ~70%

storage temp.

−20°C

Application

Used to label κ opioid receptors; precursor for the production of labeled DAKLI; can be coupled to 125I-Bolton-Hunter reagent, FITC, biotin, etc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Goldstein et al.
Proceedings of the National Academy of Sciences of the United States of America, 85(19), 7375-7379 (1988-10-01)
We describe a synthetic ligand, "DAKLI" (Dynorphin A-analogue Kappa LIgand), related to the opioid peptide dynorphin A. A single reactive amino group at the extended carboxyl terminus permits various reporter groups to be attached, such as 125I-labeled Bolton-Hunter reagent, fluorescein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service