B4752
5-Bromocytosine
Synonym(s):
4-Amino-5-bromo-2-hydroxypyrimidine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H4BrN3O
CAS Number:
Molecular Weight:
190.00
Beilstein:
127286
EC Number:
MDL number:
UNSPSC Code:
12352204
Recommended Products
mp
240-243 °C (dec.) (lit.)
SMILES string
NC1=NC(=O)NC=C1Br
InChI
1S/C4H4BrN3O/c5-2-1-7-4(9)8-3(2)6/h1H,(H3,6,7,8,9)
InChI key
QFVKLKDEXOWFSL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M Ortiz-Lombardía et al.
Nature structural biology, 6(10), 913-917 (1999-10-03)
DNA recombination is a universal biological event responsible both for the generation of genetic diversity and for the maintenance of genome integrity. A four-way DNA junction, also termed Holliday junction, is the key intermediate in nearly all recombination processes. This
Kristin A Seiberling et al.
International forum of allergy & rhinology, 2(1), 80-84 (2012-02-09)
One theory for the pathogenesis of chronic rhinosinusitis with nasal polyps (CRSwNP) involves aberration in the expression of genes that maintain the sinonasal innate immune system. We propose that the alteration in gene expression seen in CRSwNP is a result
Rodrigo Varas et al.
Brain research, 1072(1), 72-78 (2006-01-13)
Petrosal ganglion neurons are depolarized and fire action potentials in response to acetylcholine and nicotine. However, little is known about the subtype(s) of nicotinic acetylcholine receptors involved, although alpha4 and alpha7 subunits have been identified in petrosal ganglion neurons. Cytisine
C M Nunn
Journal of biomolecular structure & dynamics, 14(1), 49-55 (1996-08-01)
The crystal structure of the DNA decamer d(AGGBrCATGCCT) has been determined to a resolution of 2.0 A and R factor of 19.6%. Crystals of d(AGGBrCATGCCT) were grown in the presence of cobalt hexammine but no cobalt hexammine ions are bound
B Lakatos et al.
FEMS microbiology letters, 171(2), 161-165 (1999-03-17)
The transport of radioactively labelled uracil into submerged mycelium of T. viride was measured by means of a membrane filtration technique. It was found to be time-dependent (up to 90 min) and concentration-dependent (up to 8 mmol l-1). Its concentration
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service