Skip to Content
Merck

Skip To

B4526

5-Bromo-4-chloro-3-indolyl α-D-mannopyranoside

≥93%

Synonym(s):

X-Man

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 464,00

About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
NACRES:
NA.83
PubChem Substance ID:
UNSPSC Code:
12352204
MDL number:

€ 464,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥93%

form

powder

solubility

DMF: 20 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](Oc2c[nH]c3ccc(Br)c(Cl)c23)[C@@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C14H15BrClNO6/c15-5-1-2-6-9(10(5)16)7(3-17-6)22-14-13(21)12(20)11(19)8(4-18)23-14/h1-3,8,11-14,17-21H,4H2/t8-,11-,12+,13+,14+/m1/s1

InChI key

OPIFSICVWOWJMJ-HAAGFXOZSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B4527B425244687
assay

≥93%

assay

≥97%

assay

≥98%

assay

≥98.0% (HPLC)

solubility

DMF: 20 mg/mL, clear, colorless to light yellow

solubility

DMF: 50 mg/mL, clear, colorless to faintly yellow

solubility

DMF: methanol (1:1): 50 mg/mL

solubility

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters