Skip to Content
Merck
All Photos(1)

Key Documents

B3561

Sigma-Aldrich

BAY 73-6691

≥98% (HPLC), powder

Synonym(s):

1-(2-Chlorophenyl)-6-[(2R)-3,3,3-trifluoro-2-methylpropyl]-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidine-4-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€222.00
25 MG
€736.00

€222.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€222.00
25 MG
€736.00

About This Item

Empirical Formula (Hill Notation):
C15H12ClF3N4O
CAS Number:
Molecular Weight:
356.73
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

€222.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

DMSO: >20 mg/mL

originator

Bayer

storage temp.

2-8°C

SMILES string

C[C@H](CC1=Nc2c(cnn2-c3ccccc3Cl)C(=O)N1)C(F)(F)F

InChI

1S/C15H12ClF3N4O/c1-8(15(17,18)19)6-12-21-13-9(14(24)22-12)7-20-23(13)11-5-3-2-4-10(11)16/h2-5,7-8H,6H2,1H3,(H,21,22,24)/t8-/m1/s1

InChI key

FFPXPXOAFQCNBS-MRVPVSSYSA-N

Application

BAY 73-6691 has been used as a phosphodiesterase 9 inhibitor in ischemic heart samples[1] and in structural studies.[2] It may be used as a PDE 9 inhibitor in neuroblastoma SH-SY5Y cells.[3]

Biochem/physiol Actions

BAY 73-6691 is the first potent, selective, cell permeable inhibitor of phosphodiesterase 9 (IC50 for human PDE9 = 55 nM; little to no activity at other PDEs); PDE9 is highly selective for cGMP and is detected in brain, kidney, spleen, prostate, colon, and intestine.
BAY 73-6691 was characterized in vitro as the first potent and selective inhibitor of phosphodiesterase 9 (PDE9), which is currently under preclinical development for the treatment of Alzheimer′s disease. This compound selectively inhibits human (IC50 = 55 nM) and murine (IC50 = 100 nM) PDE9 activity in vitro and shows only moderate activity against other cyclic nucleotide-specific phosphodiesterases. BAY 73-6691 alone did not significantly increase basal cGMP levels. The PDE9 inhibitor significantly potentiated the cGMP signals generated by sGC activating compounds such as BAY 58-2667 or 5-cyclopropyl-2-[1-(2-fluorobenzyl)-1H-pyrazolo[3,4-b]pyridin-3-yl]pyrimidin-4-ylamine (BAY 41-2272) and induced leftward shifts of the corresponding concentration-response curves. The newly generated PDE9 reporter cell line show that BAY 73-6691 is able to efficiently penetrate cells and to inhibit intracellular PDE9 activity.†

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphodiesterases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insight into binding of phosphodiesterase-9A selective inhibitors by crystal structures and mutagenesis
Wang H, et al.
Journal of Medicinal Chemistry, 53(4), 1726-1731 (2010)
Protective effects of BAY 73-6691, a selective inhibitor of phosphodiesterase 9, on amyloid-beta peptides-induced oxidative stress in in-vivo and in-vitro models of Alzheimer's disease
Li J, et al.
Brain Research, 1642, 327-335 (2016)
Maritza Cavalcante Barbosa et al.
Basic & clinical pharmacology & toxicology, 118(4), 271-278 (2015-09-09)
The aim of the study was to investigate the possible anti-inflammatory and antioxidant effects of BAY 73-6691 on neutrophils from SCA patients. This study included 35 patients with a molecular diagnosis of SCA, whose neutrophils were isolated and treated with
sGC-cGMP-PKG pathway stimulation protects the healthy but not the failing right ventricle of rats against ischemia and reperfusion injury
Andersen A, et al.
International Journal of Cardiology, 223, 674-680 (2016)
Dewi Safitri et al.
Biochemical pharmacology, 174, 113823-113823 (2020-01-29)
Supressed levels of intracellular cAMP have been associated with malignancy. Thus, elevating cAMP through activation of adenylyl cyclase (AC) or by inhibition of phosphodiesterase (PDE) may be therapeutically beneficial. Here, we demonstrate that elevated cAMP levels suppress growth in C6

Articles

Cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and/or cGMP. There are 11 different mammalian PDE families.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service