Skip to Content
Merck

Skip To

B3188

BYK204165

≥98% (HPLC)

Synonym(s):

4-(1-methyl-1H-pyrrol-2-ylmethylene)-4H-isoquinolin-1,3-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 162,00

About This Item

Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated

€ 162,00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

yellow

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

Cn1cccc1\C=C2/C(=O)NC(=O)c3ccccc23

InChI

1S/C15H12N2O2/c1-17-8-4-5-10(17)9-13-11-6-2-3-7-12(11)14(18)16-15(13)19/h2-9H,1H3,(H,16,18,19)/b13-9-

InChI key

BTYSIDSTHDDAJW-LCYFTJDESA-N

Gene Information

human ... PARP1(142)
mouse ... PARP1(11545)
rat ... PARP1(25591)

Biochem/physiol Actions

BYK204165 is a potent and selective inhibitor of poly(ADP-ribose) polymerase 1 (PARP1). It is 100-fold selective for PARP-1 over PARP-2.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
528824AB16661SRP0192
description

≥98% (HPLC)

description

The PARP Inhibitor XIV, also referenced under CAS 1104546-89-5, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.

description

serum, Chemicon®

description

recombinant, expressed in baculovirus infected insect cells, ≥80% (SDS-PAGE)

assay

≥98% (HPLC)

assay

≥97% (sum of two isomers, HPLC)

assay

-

assay

≥80% (SDS-PAGE)

form

powder

form

solid

form

-

form

aqueous solution

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

−70°C

solubility

DMSO: >20 mg/mL

solubility

DMSO: 10 mg/mL

solubility

-

solubility

-

storage condition

desiccated

storage condition

OK to freeze, protect from light

storage condition

-

storage condition

avoid repeated freeze/thaw cycles


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters