Skip to Content
Merck
All Photos(1)

Key Documents

B2876

Sigma-Aldrich

O-Benzyl-DL-serine

Synonym(s):

(±)-2-Amino-3-benzyloxypropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCH2CH(NH2)COOH
CAS Number:
Molecular Weight:
195.22
Beilstein:
3204905
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

NC(COCc1ccccc1)C(O)=O

InChI

1S/C10H13NO3/c11-9(10(12)13)7-14-6-8-4-2-1-3-5-8/h1-5,9H,6-7,11H2,(H,12,13)

InChI key

IDGQXGPQOGUGIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service