Skip to Content
Merck
All Photos(1)

Documents

B2522

Sigma-Aldrich

Boc-Glu-OtBu

>98% (TLC)

Synonym(s):

Boc-L-glutamic acid 1-tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H25NO6
CAS Number:
Molecular Weight:
303.35
Beilstein:
3653769
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Boc-Glu-OtBu,

Assay

>98% (TLC)

form

powder

color

white

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)N[C@@H](CCC(O)=O)C(=O)OC(C)(C)C

InChI

1S/C14H25NO6/c1-13(2,3)20-11(18)9(7-8-10(16)17)15-12(19)21-14(4,5)6/h9H,7-8H2,1-6H3,(H,15,19)(H,16,17)/t9-/m0/s1

InChI key

YMOYURYWGUWMFM-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Boc-Glu-OtBu is an N-terminal protected amino acid used in solid-phase peptide synthesis (SPPS) to make unique peptides containing glutamate tert-butyl ester residues.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Shi et al.
Chemical science, 8(9), 6188-6195 (2017-10-11)
As a sudden inflammation of the pancreas, acute pancreatitis presents severe complications and a high mortality rate, despite treatment. Lipase in serum serves as an essential biomarker of acute pancreatitis and even pancreatic cancer. Therefore, developing robust, convenient and sensitive
Yang Zhang et al.
ACS applied materials & interfaces, 11(31), 27529-27535 (2019-07-11)
Characterizing over-expressed enzymes or biomarkers in living cells is critical for the molecular understanding of disease pathology and consequently for designing precision medicines. Herein, a "switch-on" probe is designed to selectively detect γ-glutamyl transpeptidase (GGT) in living cells via a
Wanyi Tai et al.
Biomaterials, 35(25), 7194-7203 (2014-05-31)
A graft copolymer with pendant drug segments can fold into nanostructures in a protein folding-like manner. The graft copolymer is constructed by directly polymerizing γ-camptothecin-glutamate N-carboxyanhydride (Glu(CPT)-NCA) on multiple sites of poly(ethylene glycol) (PEG)-based main chain via the ring open

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service