Skip to Content
Merck

Skip To

B1680

Bradyzide di(trifluoroacetate) salt hydrate

≥98%

Synonym(s):

(S)-1-[4-(4-Benzhydrylthiosemicarbazido)-3-nitrobenzenesulfonyl]-pyrrolidine-2-carboxylic acid [2-[(2-dimethylaminoethyl)methylamino]ethyl] amide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 1.140,00

About This Item

Empirical Formula (Hill Notation):
C32H42N8O5S2 · 2C2HF3O2 · xH2O
Molecular Weight:
910.90 (anhydrous basis)
UNSPSC Code:
41106200
NACRES:
NA.32
MDL number:
Assay:
≥98%
Form:
solid

€ 1.140,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98%

form

solid

solubility

H2O: >40 mg/mL

storage temp.

−20°C

Biochem/physiol Actions

Potent, orally active, non-peptide B2 bradykinin receptor antagonist.

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S1069SML2774S5451
form

solid

form

solid

form

powder

form

solid

assay

≥98%

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

room temp

solubility

H2O: >40 mg/mL

solubility

DMSO: >5 mg/mL, H2O: insoluble

solubility

H2O: 2 mg/mL, clear

solubility

deionized water: ≥5 mg/mL


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters