Skip to Content
Merck

B162

B-HT 920 dihydrochloride

solid, ≥98% (HPLC)

Synonym(s):

5,6,7,8-Tetrahydro-6-(2-propenyl)-4H-thiazolo[4,5-d]azepine-2-amine dihydrochloride, Talipexole

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C10H15N3S · 2HCl
CAS Number:
Molecular Weight:
282.23
UNSPSC Code:
12352202
PubChem Substance ID:
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

B-HT 920 dihydrochloride, solid, ≥98% (HPLC)

InChI

1S/C10H15N3S.2ClH/c1-2-5-13-6-3-8-9(4-7-13)14-10(11)12-8;;/h2H,1,3-7H2,(H2,11,12);2*1H

SMILES string

Cl.Cl.Nc1nc2CCN(CCc2s1)CC=C

InChI key

DPQAXNSOFFYKDS-UHFFFAOYSA-N

assay

≥98% (HPLC)

form

solid

color

off-white

solubility

H2O: >20 mg/mL

Gene Information

rat ... Drd2(24318)

Biochem/physiol Actions

α2-adrenoceptor agonist; D2 dopamine receptor agonist.

Features and Benefits

This compound is featured on the α2-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshitoki Takagawa et al.
Hypertension research : official journal of the Japanese Society of Hypertension, 25(2), 197-202 (2002-06-06)
To investigate the vascular endothelial dysfunction in the insulin resistance syndrome, muscarinic and alpha2-adrenergic mediated relaxations were studied in the fructose-fed rat. Male Sprague-Dawley rats were fed either fructose-rich chow (FFR, n=14) or normal chow (CNT, n=13) for 8 weeks.
Minori Enomoto et al.
Movement disorders : official journal of the Movement Disorder Society, 23(6), 811-816 (2007-12-13)
This study was done to identify the clinical characteristics of uremic restless legs syndrome (RLS). Consecutive uremic RLS patients (n = 15) and idiopathic RLS patients (iRLS; n = 20) were evaluated. The groups were compared with respect to their
M R Corboz et al.
Pulmonary pharmacology & therapeutics, 20(2), 149-156 (2006-07-01)
Nasal congestion, one of the major disease features of rhinitis, is induced by the filling of venous sinusoids causing mucosal engorgement with resultant obstruction of nasal airflow. The only available drugs that directly target the underlying vascular features driving nasal
Beatriz Civantos et al.
European journal of pharmacology, 489(1-2), 101-110 (2004-04-06)
It has been suggested that the combination of dietary Ca and Ca2+ channel antagonists could have a synergic antihypertensive effect. In this study, 3-week-old male spontaneously hypertensive rats (SHR) were randomized into four groups of animals. Two of these groups
R C Miller et al.
British journal of pharmacology, 83(4), 903-908 (1984-12-01)
In the presence of endothelium maximal contractions of rat aorta preparations evoked by B-HT 920 were about 10% of those evoked in the absence of endothelium. 6-Allyl-2-amino-5,6,7,8-tetrahydro-4H-thiazolo-(4,5-d)azepin dihydrochloride (B-HT 920, 0.1 microM to 0.1 mM) induced concentration-dependent contractions of rat

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service