Skip to Content

B161

B-HT 933 dihydrochloride

≥98% (HPLC), solid

Synonym(s):

6-Ethyl-5,6,7,8-tetrahydro-4H-oxazolo[4,5-d]azepin-2-amine dihydrochloride, Azepexole dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H15N3O · 2HCl
CAS Number:
Molecular Weight:
254.16
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

H2O: >20 mg/mL

SMILES string

Cl.Cl.CCN1CCc2nc(N)oc2CC1

InChI

1S/C9H15N3O.2ClH/c1-2-12-5-3-7-8(4-6-12)13-9(10)11-7;;/h2-6H2,1H3,(H2,10,11);2*1H

InChI key

HBLPYIOKPJVFQW-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Selective α2-adrenoceptor agonist.

Features and Benefits

This compound is featured on the α2-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.


Storage Class

13 - Non Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library