Skip to Content
Merck
All Photos(1)

Key Documents

B121

Sigma-Aldrich

Bromoacetylcholine bromide

Synonym(s):

2-(2-Bromoacetyloxy)-N.,N,N-trimethylethanaminium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€385.00

€385.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
500 MG
€385.00

About This Item

Empirical Formula (Hill Notation):
C7H15BrNO2 · Br
CAS Number:
Molecular Weight:
305.01
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€385.00


Estimated to ship on22 May 2025


Request a Bulk Order

form

powder

solubility

H2O: soluble

storage temp.

−20°C

SMILES string

[Br-].C[N+](C)(C)CCOC(=O)CBr

InChI

1S/C7H15BrNO2.BrH/c1-9(2,3)4-5-11-7(10)6-8;/h4-6H2,1-3H3;1H/q+1;/p-1

InChI key

AFIIOPIEVQSYHH-UHFFFAOYSA-M

Application

Used as an affinity alkylating agent for nicotinic receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A K L Freitas et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 53(5), e9211-e9211 (2020-04-23)
Strenuous exercise triggers deleterious effects on the intestinal epithelium, but their mechanisms are still uncertain. Here, we investigated whether a prolonged training and an additional exhaustive training protocol alter intestinal permeability and the putative effect of alanyl-glutamine (AG) pretreatment in
Acetylcholinesterase hydrolysis of halogen substituted acetylcholines.
C Y Chiou et al.
Biochemical pharmacology, 17(5), 805-815 (1968-05-01)
S Tucek
The Journal of physiology, 322, 53-69 (1982-01-01)
1. The synthesis of acetylcholine (ACh) has been measured in homogenates of the sciatic nerve, normal and denervated extensor digitorum longus (e.d.l.) muscles, and central (innervated) and peripheral (non-innervated) parts of the diaphragm of the rat. The synthesis proceeded under
Attila Rutai et al.
Shock (Augusta, Ga.), 54(1), 87-95 (2019-07-19)
The hypoxia-sensitive endothelin (ET) system plays an important role in circulatory regulation through vasoconstrictor ETA and ETB2 and vasodilator ETB1 receptors. Sepsis progression is associated with microcirculatory and mitochondrial disturbances along with tissue hypoxia. Our aim was to investigate the
Darlan Henrique Canei et al.
Veterinary world, 14(8), 2002-2008 (2021-09-28)
Urethral obstruction (UO) is a common condition in feline medicine. Severe acid-base and electrolyte disorders promote relevant electrocardiographic changes in these animals. Cardiac biomarkers such as cardiac troponin I have been shown to be useful in identifying cats with myocardial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service