Skip to Content
Merck
All Photos(1)

Key Documents

AXYVT250RS

Pipette tips for Velocity, robotic

size 250 μL, pkg of 5x960tips/cs, racked, clear, sterile

Synonym(s):

Pipette tips for Velocity, robotic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
250 μL
sterility:
sterile
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc VT-250-R-S
Corning
Pricing and availability is not currently available.

sterility

sterile

manufacturer/tradename

Axygen Inc VT-250-R-S
Corning

packaging

pkg of 5x960tips/cs

size

250 μL

volume

250 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Product is only available in China.

Other Notes

Browse all pipette tips

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service