AXYTF1000LRS
MAXYMum Recovery® pipette filter tips
racked, size 1000 μL, pkg of 5x1000tips/cs, sterile
Synonym(s):
Pipette tips, pipette tips
About This Item
RNase free
barrier
endotoxin free
filter
Corning
Recommended Products
material
clear
sterility
sterile
feature
DNA free
RNase free
barrier
endotoxin free
filter
manufacturer/tradename
Axygen Inc TF-1000-L-R-S
Corning
packaging
pkg of 5x1000tips/cs
size
1000 μL
Looking for similar products? Visit Product Comparison Guide
Application
Features and Benefits
- Tips are sterile and are stored in sealed-bottom racks with lids
- The filter creates a barrier within the tip to hinder and block aerosols, preventing cross-contamination between samples
- To prevent nuclease contamination, each polyethylene filter is robotically put into the tip and is devoid of DNA inhibitors
- For extremely low retention and high productivity, Maxymum recovery technology is used
- Nonpyrogenic
- RNase-/DNase-free
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service