Skip to Content
Merck
All Photos(1)

Key Documents

AXYR96PCRFY

Corning® Axygen® 96 Well PCR Storage Rack

size 96 wells, fluorescent yellow, pkg of 5x5racks/cs

Synonym(s):

Storage racks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.15
Pricing and availability is not currently available.

manufacturer/tradename

Axygen Inc R-96-PCR-FY
Corning

packaging

pkg of 5x5racks/cs

size

96 wells

color

fluorescent yellow

Looking for similar products? Visit Product Comparison Guide

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service