AXYPCR384Y
Corning® Axygen® PCR Microplate
size 384 wells, conical bottom yellow polypropylene wells, pkg of 5x10plates/cs
Synonym(s):
384 multiwell plate
About This Item
material
conical bottom yellow polypropylene wells
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-384-Y
Corning
packaging
pkg of 5x10plates/cs
maximum volume
40 μL
size
384 wells
working volume
40 μL
suitability
suitable for PCR
binding type
non-treated surface
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)