Skip to Content
Merck
All Photos(1)

Key Documents

AXYP96450VB

V-bottom assay plate

size 96 wells, SBS FootPrint, V-bottom blue polypropylene wells, blue, maximum volume 500 μL, pkg of 10x5plates/cs

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
V-bottom blue polypropylene wells
blue plate
size:
96 wells
Pricing and availability is not currently available.

material

V-bottom blue polypropylene wells
blue plate

sterility

non-sterile

manufacturer/tradename

Axygen Inc P-96-450V-B
Corning

packaging

pkg of 10x5plates/cs

maximum volume

500 μL

size

96 wells

working volume

450 μL

color

blue

suitability

suitable for PCR

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service