Skip to Content
Merck
All Photos(3)

Documents

AV54299

Sigma-Aldrich

Anti-GSN antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp313L0718, Anti-Gelsolin (aMyloidosis, Finnish type)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

83 kDa

species reactivity

bovine, sheep, horse, guinea pig, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GSN(2934)

Immunogen

Synthetic peptide directed towards the C terminal region of human GSN

Application

Anti-GSN antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

GSN gene encodes a 93,000-dalton actin-modulating protein localized in the cytoplasm of cells. It plays a pivotal role in regulating actin filament length as well as prevents monomer exchange by blocking or capping the "plus" ends of actin monomers and filaments. Defects in GSN gene leads to familial amyloidosis Finnish type (FAF).

Sequence

Synthetic peptide located within the following region: KPMIIYKGGTSREGGQTAPASTRLFQVRANSAGATRAVEVLPKAGALNSN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Kwiatkowski et al.
Nature, 323(6087), 455-458 (1986-10-02)
Gelsolin is representative of a class of actin-modulating proteins found in lower eukaryotes to mammals, which sever actin filaments. Gelsolin found in the cytoplasm of cells is functionally similar to a mammalian plasma protein of similar size, originally called ADF
A de la Chapelle et al.
Nature genetics, 2(2), 157-160 (1992-10-01)
Dominantly inherited familial amyloidosis, Finnish type (FAF) is caused by the accumulation of a 71-amino acid amyloidogenic fragment of mutant gelsolin (GSN). FAF is common in Finland but is very rare elsewhere. In Finland and in two American families, the
D Wen et al.
Biochemistry, 35(30), 9700-9709 (1996-07-30)
Gelsolin is a widely distributed actin binding protein that regulates actin filament length. It exists in both an intracellular and an extracellular form that is derived from a single gene by alternative splicing. Both forms contain the six homologous domains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service