Skip to Content
Merck
All Photos(1)

Key Documents

AV53597

Sigma-Aldrich

Anti-INHA antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Inhibin, α

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€423.00

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€423.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40 kDa

species reactivity

bovine, goat, sheep, human, pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... INHA(3623)

General description

NHA (inhibin, alpha) gene encodes the alpha subunit of inhibins A and B protein complexes that belongs to TGF-beta family. It is localized in the syncytiotrophoblast and the intermediate trophoblast of the placenta and may play a role in the mechanism of labor. INHA is also known as Inhibin α. Activin and inhibin are two closely related protein complexes involved in follicle-stimulating hormone (FSH) synthesis. They are produced in the gonads, pituitary gland, placenta, corpus luteum and other organs.

Immunogen

Synthetic peptide directed towards the N terminal region of human INHA

Application

Anti-INHA antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

Inhibins facilitates the regulation of numerous cellular processes including cell proliferation, apoptosis, immune response and hormone secretion. Loss of expression of inhibinα results in high grade prostate cancer. Follicle-stimulating hormone (FSH) stimulates the secretion of inhibin from the granulosa cells of the ovarian follicles in the ovaries. In turn, inhibin suppresses FSH. Inhibin secretion is diminished by GHRH, and enhanced by insulin-like growth factor-1. Inhibin may involve competing with activin for binding to activin receptors and binding to inhibin-specific receptors. Activin, inhibin and follistatin participate as intraovarian regulatory molecules involved in follicular cell proliferation, differentiation, steroidogenesis, oocyte maturation and corpus luteum function.

Sequence

Synthetic peptide located within the following region: GLAQEAEEGLFRYMFRPSQHTRSRQVTSAQLWFHTGLDRQGTAASNSSEP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G M Lambert-Messerlian et al.
Molecular and cellular endocrinology, 225(1-2), 101-108 (2004-09-29)
To date, the only routine clinical application of inhibin or activin measurement in testing for fetal abnormalities has been the use of inhibin A in prenatal screening for trisomy 21 (Down syndrome). Second trimester maternal serum levels of inhibin A
Matías L Stangaferro et al.
Animal reproduction science, 148(3-4), 97-108 (2014-07-09)
Cystic ovarian disease (COD) is an important cause of infertility in dairy cattle. Although many researchers have focused their work on the endocrine changes related to this disease, evidence indicates that intraovarian components play an important role in follicular persistence.
P van Zonneveld et al.
Human reproduction (Oxford, England), 18(3), 495-501 (2003-03-05)
The cause of declining fertility with age, in women who still have regular menstrual cycles, is not clear. Follicle development, endometrial growth and hormonal patterns were evaluated in cycles of older women (aged 41-46 years; n = 26) who previously
Stefano Luisi et al.
Human reproduction update, 11(2), 123-135 (2004-12-25)
A great deal of new information has arisen in the recent years concerning inhibin physiology and clinical relevance in reproductive medicine. It is now recognized that the two inhibin isoforms, named inhibin A and inhibin B, are produced by the
A Kondi-Pafiti et al.
Clinical and experimental obstetrics & gynecology, 40(1), 109-112 (2013-06-04)
The aim of the study was to examine, by an immunohistochemical method, the distribution of Inhibin-A and -B, in placentas from normal and pathological gestations. Sixty-two specimens of placental tissue were examined: i) ten cases from early gestations, ii) 28

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service