Skip to Content
Merck
All Photos(2)

Key Documents

AV51319

Sigma-Aldrich

Anti-GP1BA antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BSS, Anti-CD42B, Anti-CD42b-α+F12935, Anti-GP1B, Anti-Glycoprotein Ib (platelet), α polypeptide, Anti-MGC34595

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€498.00

€498.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€498.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€498.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

68 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GP1BA(2811)

Immunogen

Synthetic peptide directed towards the C terminal region of human GP1BA

Application

Anti-GP1BA antibody produced in rabbit is suitable for western blotting at a concentration of 0.25μg/ml.

Biochem/physiol Actions

Glycoprotein Ib (GP1BA) is a membrane glycoprotein present on the surface of platelets. It acts as the receptors for von Willebrand factor that facilitates the adhesion of platelets to vascular subendothelium after vascular injury, thrombosis, platelet activation and hemostasis. Mutations in GP1BA are associated with Bernard-Soulier syndromes and platelet-type von Willebrand disease.

Sequence

Synthetic peptide located within the following region: RGSLPTFRSSLFLWVRPNGRVGPLVAGRRPSALSQGRGQDLLSTVSIRYS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maha Othman et al.
Seminars in thrombosis and hemostasis, 39(6), 663-673 (2013-08-13)
Compared with coagulation factor defects, little attention is given to defects of platelet function as causes of rare bleeding disorders. Platelet-type von Willebrand disease (PT-VWD) is an autosomal dominant bleeding disorder and is unique among platelet disorders because it is
Naomasa Yamamoto et al.
Thrombosis research, 131(4), e160-e167 (2013-02-19)
A defective platelet glycoprotein (GP) Ib/IX/V complex [von Willebrand factor (VWF) receptor] results in Bernard-Soulier syndrome (BSS), which is characterized by macrothrombocytopenia and impaired ristocetin- and thrombin-induced platelet aggregation. We found 2 independent BSS-variant families: Case I [compound heterozygous mutations
Sayer I Al-Azzam et al.
Gene, 526(2), 118-121 (2013-05-22)
Aspirin is an antiplatelet agent commonly used in treatment of patients with high risk to develop stroke and myocardial infarction. However, inter-individual variability regarding the inhibition of platelet function by aspirin is well documented. In this study, the correlation between

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service