Skip to Content
Merck
All Photos(2)

Key Documents

AV49316

Sigma-Aldrich

Anti-TEX2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp781G0721, Anti-HT008, Anti-KIAA1738, Anti-TMEM96, Anti-Testis expressed 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€374.00

€374.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€374.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€374.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

126 kDa

species reactivity

rat, guinea pig, bovine, rabbit, mouse, human, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TEX2(55852)

Immunogen

Synthetic peptide directed towards the N terminal region of human TEX2

Application

Anti-TEX2 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5μg/ml.

Biochem/physiol Actions

Testis expressed 2 (TEX2; HT008) is a chaperone protein containing SMP domain that regulates sphingolipid metabolism.

Sequence

Synthetic peptide located within the following region: KSLSTEVEPKESPHPARHRHLMKTLVKSLSTDTSRQESDTVSYKPPDSKL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ian Lee et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 20(2), 202-206 (2006-02-02)
We have analyzed the sequence of a mitochondrial integral membrane protein, Mdm12, and found that it forms the prototype for a novel domain, designated the SMP domain, that is common to an extended family of membrane-associated proteins. Comprehensive sequence searches

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service