Skip to Content
Merck
All Photos(1)

Key Documents

AV49036

Sigma-Aldrich

Anti-GSTZ1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-GSTZ1-1, Anti-Gutathione transferase zeta 1 (maleylacetoacetate isomerase), Anti-MAAI, Anti-MAI, Anti-MGC2029

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€289.00

€289.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€289.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€289.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

24 kDa

species reactivity

mouse, pig, bovine, human, dog, rabbit, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GSTZ1(2954)

Immunogen

Synthetic peptide directed towards the N terminal region of human GSTZ1

Application

Anti-GSTZ1 antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml.

Biochem/physiol Actions

Glutathione S-transferase zeta 1 (GSTZ1) belongs to glutathione S-transferase (GSTs) super-family involved in detoxification of carcinogens and drugs by conjugation with glutathione. GSTZ1 is also involved in the metabolism of phenylalanine and tyrosine. Defects in GSTZ1 gene result in metabolic disorders such as phenylketonuria, alkaptonuria and tyrosinaemia.

Sequence

Synthetic peptide located within the following region: MQAGKPILYSYFRSSCSWRVRIALALKGIDYETVPINLIKDGGQQFSKDF

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Polekhina et al.
Biochemistry, 40(6), 1567-1576 (2001-05-01)
Maleylacetoacetate isomerase (MAAI), a key enzyme in the metabolic degradation of phenylalanine and tyrosine, catalyzes the glutathione-dependent isomerization of maleylacetoacetate to fumarylacetoacetate. Deficiencies in enzymes along the degradation pathway lead to serious diseases including phenylketonuria, alkaptonuria, and the fatal disease
Li Yin et al.
Journal of molecular recognition : JMR, 26(1), 38-45 (2013-01-03)
Accumulating evidence shows that glutathione peroxidase (GPX, EC.1.11.1.9), one of the most important antioxidant selenoenzymes, plays an essential role in protecting cells and tissues against oxidative damage by catalyzing the reduction of hydrogen peroxide by glutathione. Unfortunately, because of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service