Skip to Content
Merck
All Photos(1)

Key Documents

AV48130

Sigma-Aldrich

Anti-ALDOA antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-ALDA, Anti-Aldolase A, fructose-bisphosphate, Anti-MGC10942, Anti-MGC17716, Anti-MGC17767

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€289.00

€289.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€289.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€289.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

dog, human, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALDOA(226)

General description

Aldolase A, fructose-bisphosphate (ALDOA) is a ubiquitous glycolytic enzyme expressed in developing embryo and in adult muscle. It is involved in a wide range of cellular functions such as maintenance of striated muscle contraction, cell shape and mobility, actin filament organization and ATP biosynthetic process.[1][2]

Immunogen

Synthetic peptide directed towards the N terminal region of human ALDOA

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Rabbit Anti-ALDOA antibody has been used for western blot applications at a dilution of 1:1000.

Biochem/physiol Actions

In glycolysis, ALDOA (Aldolase A, fructose-bisphosphate) catalyzes the reversible reaction of fructose-1,6-bisphosphate to glyceraldehydes-3-phosphate and dihydroxyacetone phosphate. High expression level of ALDOA is reported in various forms of malignant cancers, including human lung squamous, renal cell and hepatocellular carcinomas.[2] ALDOA deficiency causes myopathy and hemolytic anemia.

Sequence

Synthetic peptide located within the following region: MPYQYPALTPEQKKELSDIAHRIVAPGKGILAADESTGSIAKRLQSIGTE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan Cawood et al.
PloS one, 6(1), e16152-e16152 (2011-01-26)
MicroRNAs are small non-coding RNA molecules that regulate mRNA translation and stability by binding to complementary sequences usually within the 3' un-translated region (UTR). We have previously shown that the hepatic toxicity caused by wild-type Adenovirus 5 (Ad5WT) in mice
Brigitte Picard et al.
Journal of proteomics, 200, 1-10 (2019-03-22)
This study analyzed the effect of gender on the abundances of 20 protein biomarkers of tenderness and/or intramuscular fat content in five muscles: Longissimus thoracis, previously identified as biomarkers of tenderness and/or intramuscular Semimembranosus, Rectus abdominis, Triceps brachii and Semitendinosus
Brigitte Picard et al.
PeerJ, 6, e4891-e4891 (2018-06-13)
Tenderness and intramuscular fat content are key attributes for beef sensory qualities. Recently some proteomic analysis revealed several proteins which are considered as good biomarkers of these quality traits. This study focuses on the analysis of 20 of these proteins
S Miwa et al.
American journal of hematology, 11(4), 425-437 (1981-12-01)
Two cases of red cell aldolase deficiency associated with congenital nonspherocytic hemolytic anemia are reported. The proband is a fourteen-month-old Japanese boy. Consanguineous marriage was not proven but probable in this family, as the parents were born in the same
Mohammed Gagaoua et al.
Meat science, 145, 308-319 (2018-07-18)
Reverse Phase Protein Arrays (RPPA) were applied for the quantification and validation of protein biomarkers of beef qualities on M. longissimus thoracis sampled early post-mortem from young Charolais bulls. pHu was related to six proteins, three of which are glycolytic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service