Skip to Content
Merck
All Photos(2)

Key Documents

AV46493

Sigma-Aldrich

Anti-CCPG1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CPR8, Anti-Cell cycle progression 1, Anti-KIAA1254

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€498.00

€498.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1582


Select a Size

Change View
100 μL
€498.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€498.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1582

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

87 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCPG1(9236)

Immunogen

Synthetic peptide directed towards the middle region of human CCPG1

Biochem/physiol Actions

CCPG1 acts as an assembly platform for Rho protein signaling complexes. It limits guanine nucleotide exchange activity of MCF2L toward RHOA, which results in an inhibition of both its transcriptional activation ability and its transforming activity. It does not inhibit activity of MCF2L toward CDC42, or activity of MCF2 toward either RHOA or CDC42. It may be involved in cell cycle regulation.

Sequence

Synthetic peptide located within the following region: TNLATENQYLRVSLEKEEKALSSLQEELNKLREQIRILEDKGTSTELVKE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhanhui Wang et al.
Liver international : official journal of the International Association for the Study of the Liver, 35(4), 1403-1415 (2014-08-08)
The molecular cochaperone CDC37 regulates the activities of multiple protein kinases, and is an attractive broad-spectrum target in many types of cancers in which it is over-expressed. This study investigates the antitumour effects of inhibiting CDC37 in human hepatocellular carcinoma

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service