Skip to Content
Merck
All Photos(2)

Key Documents

AV45495

Sigma-Aldrich

Anti-CHST1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-C6ST, Anti-Carbohydrate (keratan sulfate Gal-6) sulfotransferase 1, Anti-KS6ST, Anti-KSGal6ST

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€327.00

€327.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€327.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€327.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

47 kDa

species reactivity

guinea pig, dog, mouse, bovine, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHST1(8534)

Immunogen

Synthetic peptide directed towards the N terminal region of human CHST1

Application

Anti-CHST1 antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

CHST1 is a sulfotransferase that is localized to Golgi complex. It transfers sulfate from 3′phosphoadenosine 5′phospho-sulfate to the 6-hydroxyl group of N-acetylglucosamine on proteoglycan keratin. CHST1 contributes to the generation of functional L-selectin ligands in vascular endothelial cells.

Sequence

Synthetic peptide located within the following region: SFVGQLFNQHLDVFYLFEPLYHVQNTLIPRFTQGKSPADRRVMLGASRDL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Li et al.
Journal of leukocyte biology, 69(4), 565-574 (2001-04-20)
Sulfation is an essential component of the selectin ligands, potentially mediated by members of a new family of carbohydrate sulfotransferases. In this study, we assessed the contributions of CHST1, CHST2, CHST3, and CHST4 in producing functional L-selectin ligands. Human umbilical
A Bistrup et al.
The Journal of cell biology, 145(4), 899-910 (1999-05-20)
L-selectin, a lectin-like receptor, mediates rolling of lymphocytes on high endothelial venules (HEVs) in secondary lymphoid organs by interacting with HEV ligands. These ligands consist of a complex of sialomucins, candidates for which are glycosylation- dependent cell adhesion molecule 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service